Learn More
Chemglass Life Sciences Insert, 0.25mL, Glass, Flat Bottom
Supplier: Chemglass Life Sciences CV14140531
Insert, 0.25mL, Glass, Flat Bottom. Glass inserts have been engineered to fit the opening of standard opening screw thread and standard opening crimp top vials containing a nominal I.D. of 5mm. Inserts are available assembled with polymer bottom spring or patented Top Spring™. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.