Learn More
Chemglass Life Sciences Glass Insert, 0.25mL, BM with Bottom Spring
Supplier: Chemglass Life Sciences CV14190629
Glass Insert, 0.25mL, BM with Bottom Spring. Due to the conical design of the snap plug, limited volume inserts used in the shell vial stay centered and straight allowing for easy sample removal.