Learn More
Chemglass Life Sciences Glass Insert, 0.05mL, BM with Bottom Spring
Supplier: Chemglass Life Sciences CV14020625
Glass Insert, 0.05mL, BM with Bottom Spring. Both glass and plastic inserts have been engineered to fit the large opening in vials containing a nominal I.D. of 6mm. Inserts are available assembled with a polymer bottom spring or patented Top Spring™. The polymer spring acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.