Learn More
Chemglass Life Sciences Insert, .35mL, Glass, LV, No Spring
Supplier: Chemglass Life Sciences CV14280638
Insert, .35mL, Glass, LV, No Spring. These inserts have been engineered to fit the large opening of WISP style vials. Inserts are available assembled with a polymer bottom spring which acts as a shock absorber and provides a cushion against needle contact. The conical design permits complete sample removal.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.