Learn More
Innovative Med Tech Blood Gas Sampling Syringes
For laboratory or veterinary use only
Supplier: Innovative Med Tech 3999599
This item is not returnable.
View return policy
Description
- Coated with approximately 50 I.U. balanced LI-heparin
- Single use
- Sterile
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Share a Content Correction