Learn More
Electron Microscopy Sciences INFINITY1 5.0 Megapixel USB 2.0 Microscopy Camera, CMOS, Color
Supplier: Electron Microscopy Sciences 95103
INFINITY1 CMOS USB 2.0 digital cameras are designed to be a cost-effective, versatile solution for clinical, life science, materials science and educational professionals. Features include auto white balance, full exposure control, programmable gain, sub-windowing, and region of interest. It is an excellent fit for documentation and archiving applications.
Features 2, 3, and 5 mexapixel resolution. It is perfect for documentation and archiving applications and has fast frame rates.
Sensor: CMOS
Type: Color
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.