Learn More
Electron Microscopy Sciences INFINITY4-11 10.7 Megapixel Microscopy Camera, CCD, Color
Supplier: Electron Microscopy Sciences 95114
The INFINITY4 camera series offers large format megapixel Kodak sensors for a wide field of view. The perfect choice for demanding high resolution imaging requiring excellent color rendition. Features include a 12-bit digital output, binning, progressive scan electronic shutter, full exposure control, auto white balance, programmable gain, sub-windowing and region of interest - ideal for high end ophthalmic, medical, clinical and life science applications.
Features 1 megapixel resolution, a large format sensor, superior light sensitivity with high fidelity color reproduction, low noise electronics and a F-mount lens adapter. Use for brightfield, darkfield, DIC, live cell imaging, histology, pathology, cytology, defect analysis, semiconductor Inspection, metrology, gel documentation, and moderate light fluorescence applications.
Sensor: CCD
Type: Color
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.