Learn More
Andwin Scientific Dry Block/Incubator Thermometers, Thermometer, Swell, 0-80 degrees celsius
Supplier: Andwin Scientific 89095804
Individually serialized and designed for use in all standard heating blocks and water baths. Contains red organic liquid against white back glass. Thermometers can measure temperature of varying ranges depending on model.