Learn More
STEMCELL Technologies EasySep™ Human CD11b Positive Selection and Depletion Kit, For processing: 1 x 10^9 cells
Supplier: STEMCELL Technologies 1000742
Immunomagnetic positive selection of CD11b+ cells from human peripheral blood mononuclear cells (PBMCs) or leukapheresis samples
This item is not returnable.
View return policy