Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidurea, NF, 26-28%, Spectrum™ Chemical
SDP

Catalog No. 18603431
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

I1007, 39236-46-9, C11H16N8O8

Spectrum™ Chemical Imidurea, NF is an antimicrobial preservative used in topical pharmaceutical preparations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 39236-46-9
Molecular Formula C11H16N8O8
Molecular Weight (g/mol) 388.30
InChI Key ZCTXEAQXZGPWFG-UHFFFAOYNA-N
IUPAC Name 3-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-1-[({[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoyl}amino)methyl]urea
SMILES OCN1C(NC(=O)NCNC(=O)NC2N(CO)C(=O)NC2=O)C(=O)NC1=O

Specifications

CAS Max % 100%
Ignition Residue 3%
Loss on Drying 3%
Packaging Fiber Drum
Quantity 50 kg
Percent Purity 26 to 28%
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.