Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Imidapril (hydrochloride), 98%

Catalog No. AAH3739203
Click to view available options
1 g

Chemical Identifiers

CAS 89396-94-1
Molecular Formula C20H28ClN3O6
Molecular Weight (g/mol) 441.91
MDL Number MFCD00923828
InChI Key LSLQGMMMRMDXHN-GEUPQXMHSA-N
IUPAC Name hydrogen (4S)-3-[(2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]-1-methyl-2-oxoimidazolidine-4-carboxylic acid chloride
SMILES [H+].[Cl-].CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1[C@@H](CN(C)C1=O)C(O)=O

Specifications

Quantity 1 g
Formula Weight 441.91
Percent Purity 98%
Physical Form Solid
ShelfLife 5 Years
EINECSNumber 685-551-0
RTECSNumber NI8927400
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.