Learn More
Enzo Life Sciences IL-6 High Sensitivity Controls (1 Set)
SDP
Supplier:  Enzo Life Sciences ENZKIT178CTL
Encompass_Preferred
This product is a set of quantitative controls for the determination of IL-6 concentration in biological matrices, such as serum and plasma. The concentrations have been determined using the Enzo Life Sciences IL-6 (human) high sensitivity ELISA kit (ENZ-KIT178). The controls were lyophilized in porcine serum and contain human IL-6 at a low, medium and high concentration. Alternative name: Interleukin-6. Long Term Storage: -80°C. Applications: ELISA .
Catalog No. 50-201-3858
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.