Learn More
Enzo Life Sciences IL-33 (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences ALX522098C010
Encompass_Preferred
Synonyms: Interleukin-33, NF-HEV, IL-1F11. MW: ~18kDa. UniProt ID: O95760. Source: Produced in E. coli. Untagged human IL-33 (aa 112-270). Purification: Purified by multi-step chromatography. Formulation: Lyophilized. Contains PBS.
Catalog No. 50-210-0502
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.