Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-13 Rat anti-Human, LE/AF (Low Endotoxin/Azide Free), Clone: JES10-5A2, Southern Biotech™
Rat Monoclonal Antibody
Supplier: Southern Biotechnology Associates 1012514
Description
SouthernBiotech cytokine antibodies offer high sensitivity and low background for use in a variety of applications.
Specifications
IL-13 | |
Monoclonal | |
0.5mg/mL | |
Supplied in PBS | |
COS-expressed human IL-13 | |
RUO | |
Human | |
IgG1 |
ELISA, Dot Blot, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Neutralization | |
JES10-5A2 | |
LE/AF | |
Rat | |
0.5 mg | |
Primary | |
Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction