Learn More
Proteintech Group Inc IHCeasy NeuN Ready-To-Use IHC Kit
SDP
Supplier:  Proteintech Group Inc KHC000350T
Encompass_Preferred
KHC0003 is a ready-to-use IHC kit for staining of NeuN. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Catalog No. 50-225-8485
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.