Learn More
Enzo Life Sciences IHC tissue primer (100ml)
Supplier: Enzo Life Sciences ADI9502340100
The IHC tissue primer is a universal enzyme blocking reagent to use in horse radish peroxidase (HRP) and alkaline phosphatase (AP) based immunohistochemical staining procedures done on paraffin tissue sections, frozen tissue sections and cell smears to suppress nonspecific staining due to endogenous enzyme activity. Formulation: Liquid. Tissue primer. TRIS-based alkaline solution with 30% H2O2. Long Term Storage: +4°C. Handling: Store tightly sealed in the dark. Protect from light. Applications: IHC .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.