Learn More
IgG Fc Porcine anti-Human, Polyclonal, Secondary Antibody, Novus Biologicals™
SDP
Pig Polyclonal Antibody
Supplier:  Novus Biologicals NBP269497
 View more versions of this product
Catalog No. NBP2-69497
Description
Description
IgG Fc Polyclonal specifically detects IgG Fc in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunoblotting, Dot Blot.
Specifications
Specifications
IgG Fc fragment
Polyclonal
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
Porcine
Whole Antiserum
Secondary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
ELISA, Immunocytochemistry, Immunofluorescence, Immunoblot, Dot Blot
Unconjugated
Whole antiserum with No Preservative
Highly purified Fc fragment of normal IgG isolated from pooled human serum.
1 mL
Human
Antisera
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.