Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-γ Mouse anti-Human, Rhesus Ma, Cynomolgus, Chimpanzee, Pigtail Macaque, African Green Monkey, Sooty Mangabey, R-PE, Clone: B27, Southern Biotech™
Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 1011409
Description
SouthernBiotech cytokine antibodies offer high sensitivity and low background for use in a variety of applications.
Specifications
IFN-γ | |
B27 | |
R-PE | |
Conjugated | |
E. coli-expressed human IFN-γ | |
100 tests | |
Store at 2-8°C; Protect from light; Do not freeze | |
Monoclonal |
Flow Cytometry | |
Lot specific | |
Supplied in PBS/NaN3 and a stabilizing agent | |
Mouse | |
IgG1 | |
RUO | |
Primary | |
Chimpanzee, Cynomolgus, Human, Monkey, Pigtailed Macaque, Rhesus Monkey, Sooty Mangabey |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction