Learn More
Aat Bioquest iFluor 488-Wheat Germ Agglutinin WGA Conjugate (1 mg)
SDP
Supplier:  Aat Bioquest 25530
Encompass_Preferred
Wheat germ agglutinin WGA is a lectin that binds to N-acetyl-D-glucosamine and sialic acid. This catalog is for 1 mg.
Catalog No. 50-225-0163
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.