Learn More
Abbott™ ID NOW RSV Test Kit
Highly sensitive rapid diagnostic tests for RSV are needed to help guide management in the clinical setting. Combines the benefits of speed and accuracy on an easy to use system.
$2493.00
Specifications
Clia Complexity | Waived |
---|---|
CE Marker | Yes |
Detectable Analytes | Respiratory Syncytial Virus (RSV) |
Alert:
Refer to Package Insert for list of warnings
Catalog Number | Mfr. No. | Quantity | dimType | Shelf Life | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | dimType | Shelf Life | Price | Quantity & Availability | |||
23-043-632
|
Abbott™
435000 |
24/Pk. | 4 Months at launch, will increase |
Pack of 24 for $2,493.00
|
|
||||
Description
- Rapid molecular in vitro diagnostic test utilizing isothermal Nucleic Acid Amplification Technology for the qualitative detection of Respiratory Syncytial Virus l nucleic acid in nasopharyngeal swabs or nasopharyngeal swabs
- Eluted in Viral Transport Media (VTM)
- Intended to aid in the rapid diagnosis of Respiratory Syncytial Virus (RSV)
- Delivers molecular RSV results in 13 minutes or less on the unique Abbott™ ID NOW platform.
- Allows to make real-time clinical decisions which impact patient care
Specifications
Waived | |
Yes | |
Respiratory Syncytial Virus (RSV) |
Safety and Handling
ShelfLife : 4 Months at launch, will increase