Learn More
anti-ICAM-1/CD54, Alexa Fluor 546, Clone: 117G12.02, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals DDX0150A546
Catalog No. DDX0150A546
Explore all special offers
Add to Cart
Description
Description
ICAM-1/CD54 Monoclonal antibody specifically detects ICAM-1/CD54 in Human, Sheep samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ICAM-1/CD54
Monoclonal
Alexa Fluor 546
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Mouse
Ion exchange chromatography
RUO
Primary
Human, Sheep
IgG1
Immunocytochemistry, Immunofluorescence
117G12.02
Immunocytochemistry/Immunofluorescence 1:10-1:2000
ICAM1
CD54 was used as immunogen.
0.1 mg
Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
3383
Store at -20C in the dark. Avoid freeze-thaw cycles.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only