Learn More
New England Biolabs, Inc. Hydrophilic Streptavidin Magnetic Beads – 5 ml
SDP
Supplier:  New England Biolabs, Inc. S1421S
Encompass
Hydrophilic Streptavidin Magnetic Beads are 2 micron superparamagnetic particles covalently coupled to a highly pure form of streptavidin. The beads can be used to capture biotin labeled substrates including antigens, antibodies and nucleic acids. The strength of the biotin-streptavidin interaction coupled with the low non-specific binding of streptavidin permits captured substrates to be useful as ligands in subsequent experiments including mRNA isolation and the capture of primary or secondary antibodies
Catalog No. 50-448-5
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.