Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrogen Potassium ATPase Beta Antibody (2G11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NB3005830.025ML
Description
Hydrogen Potassium ATPase Beta Monoclonal specifically detects Hydrogen Potassium ATPase Beta in Human, Mouse, Rat, Porcine, Bovine, Canine, Ferret, Rabbit samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.Specifications
Hydrogen Potassium ATPase Beta | |
Monoclonal | |
Unconjugated | |
ATP6B, ATPase, H+/K+ exchanging, beta polypeptide, ATPase, H+/K+ transporting, beta polypeptide, Gastric H(+)/K(+) ATPase subunit beta, gastric H+/K+ ATPase beta subunit, gastric hydrogen-potassium ATPase, beta, potassium-transporting ATPase beta chain, potassium-transporting ATPase subunit beta, Proton pump beta chain | |
Mouse | |
Unpurified | |
RUO | |
496 | |
Human | |
Ascites |
Western Blot | |
2G11 | |
Western Blot 1:4000, Flow Cytometry 1:50, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:2000, Immunohistochemistry-Frozen 1:2000, Dot Blot 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
ATP4B | |
Purified 34 kDa core peptide from deglycosylated hog gastric microsomes. | |
0.025 mL | |
Primary | |
Detects the beta-subunit of hydrogen/potassium ATPase. | |
Store at -20°C. Avoid freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction