Learn More
HYDRANAL™-Standard 5.0, Honeywell Fluka™
Test solution for volumetric Karl Fischer titration (water content ~5.8 mg/g, corresponds to ~5.0 mg/mL, exact value on report of analysis)
Supplier:  Honeywell Chemicals 348136X100ML/DEL
 View more versions of this product
Catalog No. 60-046-462
Description
Description
Advantages of HYDRANAL- Auxiliaries
  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HYDRANAL™-Standard 5.0
27°C (80.6°F)
UN1993C
6 x 100mL
95-47-6, 71-36-3
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
P210-P261-P280-P305 + P351 + P338 + P310-P370 + P378
DOTInformation : 3 / PGIII
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.