Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRANAL™ - Coulomat AG-Oven, Reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm, Honeywell Fluka
Supplier: Honeywell Chemicals 347396X500MLUS/DEL
Description
Specifications
HYDRANAL™ - Coulomat AG-Oven | |
64°C | |
13°C | |
≥50.0000%, ≥20.0000%, ≥10.0000%, ≥5.0000%, ≥5.0000%, ≥5.0000% | |
57-55-6, 111-42-2, 288-32-4, 7446-09-5, 68007-08-9 | |
UN1230 |
6 x 500mL | |
0.980 g/cm3 | |
Reagent for coulometric KF titration with oven (anolyte solution), for cells with and without diaphragm | |
<70.0000%, <30.0000%, <20.0000%, <10.0000%, <10.0000%, <10.0000% | |
Glass Bottle | |
Karl Fischer |
Safety and Handling
ShelfLife : 1800 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol Solution
Recommended Storage : Room Temp
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction