Learn More
Glass Expansion HydraMist Kit for Thermo 6000/7000 Series ICP-OES
Supplier: Glass Expansion KT1179
HydraMist Kit. Compatible with Thermo 6000/7000 Series ICP-OES. Nebulizer not included. The HydraMist spray chamber can simultaneously determine As, Se, Sb, and Hg using hydride generation and non-hydride forming elements using conventional pneumatic nebulization. The HydraMist spray chamber will lower the detection limits between 15 and 20-fold for the hydride forming elements in sensitive mode, and 10-fold using the simple mode, without compromising the analytical performance of the non-hydride forming elements.Replacement parts include 501907304, 70-803-1808, GAZ-08-4, 60-703-1119, 70-803-2078, 70-803-0746, 70-803-1350, 501906811, NC2046952, 501906911, NC1943061, 70-803-1796, 70-803-1797, and 60-808-1187.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.