Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva HyClone™ Newborn Calf Serum (U.S.)
Augment cell culture growth with U.S.-sourced HyClone™ Newborn Bovine Calf Serum.
Supplier: Cytiva SH30118.03IR2540
Description
- Filtered through sequential 100nm filters and sourced from animals that are typically less than 10 days old
Specifications
500 mL | |
Newborn Calf Serum (U.S.) | |
No | |
Irradiated, 25 to 40 doses |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction