Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum (New Zealand)
A cost-effective alternative to Fetal Bovine Serum (FBS).
Supplier: Cytiva SH30626.03IR
Description
Specifications
Filtered through sequential 100 nm pore-size rated filters | |
HyClone™ | |
Lactate Dehydrogenase | |
Alkaline Phosphatase | |
No | |
New Zealand | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
Iron-supplemented Newborn Bovine Calf Serum |
1 L | |
Sterile | |
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG | |
≤ 25 EU/mL | |
≤ 25 mg/dL | |
Irradiated | |
Yes | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction