Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human Wnt-9b Recombinant Protein, PeproTech®
CHO cells Recombinant Protein
Supplier: Gibco™ 12049250UG
This item is not returnable.
View return policy
Description
WNT9B, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.Specifications
O14905 | |
protein with no preservative | |
36.9 | |
2.5 x 100 μg | |
-20°C | |
<1 EU/μg | |
Wnt9b | |
Unconjugated | |
SYFGLTGREV LTPFPGLGTA AAPAQGGAHL KQCDLLKLSR RQKQLCRREP GLAETLRDAA HLGLLECQFQ FRHERWNCSL EGRMGLLKRG FKETAFLYAV SSAALTHTLA RACSAGRMER CTCDDSPGLE SRQAWQWGVC GDNLKYSTKF LSNFLGSKRG NKDLRARADA HNTHVGIKAV KSGLRTTCKC HGVSGSCAVR TCWKQLSPFR ETGQVLKLRY DSAVKVSSAT NEALGRLELW APARQGSLTK GLAPRSGDLV YMEDSPSFCR PSKYSPGTAG RVCSREASCS SLCCGRGYDT QSRLVAFSCH CQVQWCCYVE CQQCVQEELV YTCKH |
Functional Assay | |
7484 | |
Human Wnt-9 b | |
CHO cells | |
RUO | |
clf; clf1; protein Wnt-14 b; Protein Wnt-15; Protein Wnt-9 b; UNQ6973/PRO21956; wingless-type MMTV integration site 15; wingless-type MMTV integration site 9 B; wingless-type MMTV integration site family member 9 B; wingless-type MMTV integration site family, member 15; wingless-type MMTV integration site family, member 9 B; Wnt family member 9 B; Wnt14b; wnt-14 b; Wnt15; wnt-15; Wnt9b | |
Human | |
Recombinant | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction