Learn More
Proteintech Group Inc Human TNFR1 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0005196T
Encompass_Preferred
Tumor necrosis factor (TNF) is a multifunctional cytokine that plays a key role in regulating inflammation, immune functions, host defense, and apoptosis. TNF signals through two distinct cell surface receptors, TNFR1 (TNFRSF1A, CD120a) and TNFR2 (TNFRSF1B, CD120b). TNFR1, which contains a death domain (DD) within its intracytoplasmic region, is thought to be the key receptor for TNF signaling. This receptor can be released to the extracellular space by two mechanisms, ectodomain shedding and constitutive release of full-length 55-kDa TNFR1. Soluble TNFR1 (sTNFR1) could function as TNF-binding protein, inhibiting TNF bioactivity. This ELISA kit can be used to measure natural and recombinant human sTNFR1.
Catalog No. 50-197-4020
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.