Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human TGF-beta 1 Recombinant Protein, PeproTech®
CHO cells Recombinant Protein
$89.50 - $5760.00
Specifications
Accession Number | P01137 |
---|---|
For Use With (Application) | Functional Assay |
Gene ID (Entrez) | 7040 |
Molecular Weight (g/mol) | 25 |
Name | Human TGF-beta 1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
10021C100UG
|
Gibco™
10021C100UG |
100 μg |
Each for $1,154.00
|
|
|||||
10021C10UG
|
Gibco™
10021C10UG |
10 μg |
Each for $217.00
|
|
|||||
10-021-C01M
|
Gibco™
10021C01M |
2 x 500 μg |
Each for $5,760.00
|
|
|||||
10021C250UG
|
Gibco™
10021C250UG |
250 μg |
Each for $2,015.00
|
|
|||||
100-21C-2UG
|
Gibco™
10021C2UG |
2 μg |
Each for $89.50
|
|
|||||
10021C500UG
|
Gibco™
10021C500UG |
500 μg |
Each for $3,460.00
|
|
|||||
10021C50UG
|
Gibco™
10021C50UG |
50 μg |
Each for $692.00
|
|
|||||
Description
TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.Specifications
P01137 | |
7040 | |
Human TGF-beta 1 | |
-20°C | |
<1 EU/μg | |
Tgfb1 | |
Unconjugated | |
ALDTNYCFSS TEKNCCVRQL YIDFRKDLGW KWIHEPKGYH ANFCLGPCPY IWSLDTQYSK VLALYNQHNP GASAAPCCVP QALEPLPIVY YVGRKPKVEQ LSNMIVRSCK CS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
25 | |
CHO cells | |
RUO | |
CED; cytokine; DPD1; LAP; Latency-associated peptide; prepro-transforming growth factor beta-1; regulatory protein; TGF b; TGF b 1; TGF beta; tgf beta 1; tgf beta1; TGF beta-1; TGF beta-1 protein; TGF ß; TGF ß 1; TGF ß1; Tgfb; TGFB1; TGF-B1; Tgfb-1; TGFbeta; TGF-beta 1; TGF-beta 1 protein; TGFbeta1; TGF-beta1; TGF-beta-1; TGFß1; Transforming growth factor; transforming growth factor b1; transforming growth factor beta; transforming growth factor beta 1; transforming growth factor beta-1; Transforming growth factor beta-1 proprotein; transforming growth factor beta-1 proprotein; transforming growth factor beta-1; transforming growth factor ß1; transforming growth factor, beta 1; transforming growth factor, beta 1 (Camurati-Engelmann disease); transforming growth factor, beta-1; transforming growth factor-beta 1; transforming growth factor-beta I; transforming growth factor-beta-1; transforming growth factor-beta-1 precursor; unnamed protein product | |
Human | |
Recombinant | |
Lyophilized |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title