Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human SPARC Recombinant Protein, PeproTech®
CHO cells Recombinant Protein
$87.00 - $1620.00
Specifications
Accession Number | P09486 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 6678 |
Molecular Weight (g/mol) | 32.7 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
12036100UG
|
Gibco™
12036100UG |
100 μg |
Each for $324.00
|
|
|||||
120-36-10UG
|
Gibco™
1203610UG |
10 μg |
Each for $87.00
|
|
|||||
120-36-1MG
|
Gibco™
120361MG |
1 mg |
Each for $1,620.00
|
|
|||||
12036250UG
|
Gibco™
12036250UG |
250 μg |
Each for $567.00
|
|
|||||
12036500UG
|
Gibco™
12036500UG |
500 μg |
Each for $972.00
|
|
|||||
120-36-50UG
|
Gibco™
1203650UG |
50 μg |
Each for $211.00
|
|
|||||
Description
SPARC gene encodes a cysteine-rich acidic matrix-associated protein. The encoded protein is required for the collagen in bone to become calcified but is also involved in extracellular matrix synthesis and promotion of changes to cell shape. The gene product has been associated with tumor suppression but has also been correlated with metastasis based on changes to cell shape which can promote tumor cell invasion. Three transcript variants encoding different isoforms have been found for this gene.Specifications
P09486 | |
protein with no preservative | |
32.7 | |
CHO cells | |
RUO | |
Basement membrane protein 40; basement-membrane protein 40; BM40; BM-40; Cysteine rich protein; OI17; ON; Osteonectin; osteonectin precursor; Secreted acidic cystein rich glycoprotein; secreted acidic cysteine rich glycoprotein; Secreted acidic cystein-rich glycoprotein (osteonectin); secreted protein acidic and cysteine rich; secreted protein acidic and rich in cysteine; Secreted protein acidic and rich in cysteine (SPARC); secreted protein, acidic, cysteine-rich (osteonectin); secreted protein, acidic, cysteine-rich [osteonectin]; SPARC | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
6678 | |
Human SPARC | |
-20°C | |
<1 EU/μg | |
SPARC | |
Unconjugated | |
APQQEALPDE TEVVEETVAE VTEVSVGANP VQVEVGEFDD GAEETEEEVV AENPCQNHHC KHGKVCELDE NNTPMCVCQD PTSCPAPIGE FEKVCSNDNK TFDSSCHFFA TKCTLEGTKK GHKLHLDYIG PCKYIPPCLD SELTEFPLRM RDWLKNVLVT LYERDEDNNL LTEKQKLRVK KIHENEKRLE AGDHPVELLA RDFEKNYNMY IFPVHWQFGQ LDQHPIDGYL SHTELAPLRA PLIPMEHCTT RFFETCDLDN DKYIALDEWA GCFGIKQKDI DKDLVI | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title