Learn More
STEMCELL Technologies Human Recombinant DKK-1, Size: 1000 ug
SDP
Supplier:  STEMCELL Technologies 78208.2
Encompass_Preferred
Use dickkopf-related protein 1 (DKK-1) to drive cardiac and retinal differentiation from induced pluripotent stem (iPS) cells (Lian et al. Proc Natl Acad Sci USA 2012). DKK-1 is a secreted protein that inhibits the canonical WNT pathway by competitive binding to low-density lipoprotein receptors (LRP)-5 and 6 (Niehrs. Oncogene 2006). This protein regulates embryonic development (Lieven O et al. Dev Biol 2010) and has been shown to support the generation of myeloid-derived suppressor cells (MDSCs) (D’Amico et al. J Exp Med 2016). DKK-1 from thrombocytes also induces Th2 cell polarization and potentiates Th2 cell cytokine expression (Chae et al. Immunity 2016). Human recombinant DKK-1 from STEMCELL contains a His-residue tag at the carboxyl end of the polypeptide chain and comes lyophilized with ≥ 95% purity EC50 4 μg/mL endotoxin levels verified 0.2 EU/μg protein. Alternative names Dickkopf-1 Dickko
Catalog No. 50-239-0244
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.