Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human R-Spondin-2 Recombinant Protein, PeproTech®
CHO cells Recombinant Protein
$87.00 - $3240.00
Specifications
Accession Number | Q6UXX9 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 340419 |
Molecular Weight (g/mol) | 24.4 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
12043100UG
|
Gibco™
12043100UG |
100 μg |
Each for $648.00
|
|
|||||
120-43-1MG
|
Gibco™
120431MG |
1 mg |
Each for $3,240.00
|
|
|||||
120-43-20UG
|
Gibco™
1204320UG |
20 μg |
Each for $211.00
|
|
|||||
12043250UG
|
Gibco™
12043250UG |
250 μg |
Each for $1,134.00
|
|
|||||
12043500UG
|
Gibco™
12043500UG |
500 μg |
Each for $1,944.00
|
|
|||||
120-43-5UG
|
Gibco™
120435UG |
5 μg |
Each for $87.00
|
|
|||||
Description
The R-Spondin (Rspo) proteins belong to the Rspo family of Wnt modulators. Currently, the family consists of four structurally-related, secreted ligands (Rspo 1-4), all containing furin-like and thrombospondin structural domains. The Rspo proteins can interact with the Frizzled/LRP6 receptor complex in a manner that causes the stabilization, and resulting accumulation, of the intracellular signaling protein, beta-catenin. This activity effectively activates and increases the subsequent nuclear signaling of beta-catenin. R-Spondin can also bind to the previously discovered G-protein coupled receptors, LGR-4 and LGR-5. Rspo/beta-catenin signaling can act as an inducer of the transformed phenotype, and can also regulate the proliferation and differentiation of certain stem cell populations.Specifications
Q6UXX9 | |
protein with no preservative | |
24.4 | |
CHO cells | |
RUO | |
2610028F08Rik; AA673245; CRISTIN2; cristin-2; cysteine-rich and single thrombospondin domain-containing protein 2; D430027K22; ftls; hRspo2; mCristin-2; RGD1562331; Roof plate-specific spondin-2; Rspo2; R-spondin 2; R-spondin 2 homolog; R-spondin 2 homolog (Xenopus laevis); R-spondin-2; UNQ9384/PRO34209 | |
Human | |
Recombinant | |
Lyophilized |
Functional Assay | |
340419 | |
Human R-Spondin-2 | |
-20°C | |
<1 EU/μg | |
RSPO2 | |
Unconjugated | |
ASYVSNPICK GCLSCSKDNG CSRCQQKLFF FLRREGMRQY GECLHSCPSG YYGHRAPDMN RCARCRIENC DSCFSKDFCT KCKVGFYLHR GRCFDECPDG FAPLEETMEC VEGCEVGHWS EWGTCSRNNR TCGFKWGLET RTRQIVKKPV KDTILCPTIA ESRRCKMTMR HCPGGKRTPK AKEKRNKKKK RKLIERAQEQ HSVFLATDRA NQ | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title