Learn More
Sino Biological Human MSR1 ELISA Pair Set 15PP (Sino Biological)
Supplier:  Sino Biological SEK1042715
Encompass_Preferred
Each vial contains 48 ng of recombinant Human MSR1 / CD204. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 5000 pg/mL is recommended.
Catalog No. 50-237-2933
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.