Learn More
R&D Systems™ Human mGluR2 (NP_000830) VersaClone cDNA
Glutamate receptor, metabotropic 2
Supplier:  R&D Systems™ RDC0989
Catalog No. RDC0989
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Description
Description
The Human mGluR2 (NP_000830) VersaClone cDNA clone contains the complete ORF for mGluR2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The mGluR2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Reasearch Use Only; Not for Use in Humans