Learn More
Invitrogen™ Human KV1.7 (KCNA7) (aa 405-437) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP110228
Description
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability.Specifications
Q96RP8 | |
Blocking Assay, Control | |
3743 | |
100 μL | |
RUO | |
KCNA7 | |
Recombinant | |
E. coli |
≥5.0 mg/mL | |
1M urea, PBS with no preservative; pH 7.4 | |
KV1.7 (KCNA7) | |
-20° C, Avoid Freeze/Thaw Cycles | |
HAK6; Kcna7; Kcnc7; Kv1.7; potassium channel, voltage gated shaker related subfamily A, member 7; potassium voltage gated channel, shaker related subfamily, member 7; potassium voltage-gated channel subfamily A member 7; potassium voltage-gated channel, shaker-related subfamily, member 7; potassium voltage-gated channel, shaker-related subfamily, member 7; potassium voltage gated channel, shaker related subfamily, member 7; voltage-dependent potassium channel Kv1.7; voltage-gated potassium channel KCNA7; voltage-gated potassium channel subunit Kv1.7 | |
Unconjugated | |
TEGEEAGMFSHVDMQPCGPLEGKANGGLVDGEV |