Learn More
Invitrogen™ Human KCTD18 (aa 99-175) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP106812
Description
KCTD18 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include protein homooligomerization.Specifications
Q6PI47 | |
Blocking Assay, Control | |
130535 | |
100 μL | |
RUO | |
KCTD18 | |
Human | |
YPYSLSDHLANEMETYSLRSNIELKKALTDFCDSYGLVCNKPTVWVLHYLNTSGASCESRIIGVYATKTDGTDAIEK | |
Liquid |
≥5.0 mg/mL | |
1M urea, PBS with no preservative; pH 7.4 | |
KCTD18 | |
-20° C, Avoid Freeze/Thaw Cycles | |
4932411A20Rik; 6530404F10Rik; BTB/POZ domain-containing protein KCTD18; KCTD18; potassium channel tetramerisation domain containing 18; potassium channel tetramerization domain containing 18; RGD1564820; similar to potassium channel tetramerisation domain containing 18 | |
Unconjugated | |
Recombinant | |
E. coli |