Learn More
Bethyl Laboratories, Inc Human Igg Elisa Kit
Supplier: Bethyl Laboratories, Inc E88104
Human IgG ELISA Kit, 1 x pre-coated 96-well strip plate, standard, detection antibody, sample dilution buffer, HRP solution, TMB substrate, stop solution, 20X wash buffer, sealing tape