Learn More
Proteintech Group Inc Human HER2 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0005396T
Encompass_Preferred
HER2, also known as ErbB2 and Neu, is a member of the epidermal growth factor (EGF) receptor family. Overexpression of HER2 have been reported in numerous cancers, including breast, ovarian, lung, gastric, and oral cancers. It is a therapeutic target for the treatment of breast cancer and other carcinomas. HER2 is a 185-kDa transmembrane glycoprotein, the extracellular domain (ECD) of which can be cleaved from the cell surface by matrix metalloproteases and then released into blood. Elevated serum HER2 levels are reported in 23% to 80% of patients with metastatic breast cancer. Raising serum HER2 levels have been correlated with indicators of poor prognosis and a poor response to chemotherapy.
Catalog No. 50-197-4022
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.