Learn More
CELLULAR ENGINEERING TECHNOLOGIES INC Cryopreserved vial of human keratinocyte cells containing ≥ 500,000 cells
Supplier:  CELLULAR ENGINEERING TECHNOLOGIES INC CR1017500
Encompass_Preferred
Human keratinocyte cells isolated from a normal human foreskin. These cells are frozen at the first passage and exhibit robust growth characteristics in cell culture with fast doubling times and consistent morphology.Keratinocyte cells are widely used in iPSC reprogramming workflows, cell therapy development, and the study of viral infections, oncogenesis, wound healing, and skin grafting applications. These cells have a high differentiation capacity.Vial contains approximately 500,000 cells. Shipped with dry ice.
Catalog No. NC2753007
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.