Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human EG-VEGF Recombinant Protein, PeproTech®
E. coli Recombinant Protein
$87.00 - $3240.00
Specifications
Accession Number | P58294 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 84432 |
Molecular Weight (g/mol) | 9.6 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
10044100UG
|
Gibco™
10044100UG |
100 μg |
Each for $648.00
|
|
|||||
100-44-1MG
|
Gibco™
100441MG |
1 mg |
Each for $3,240.00
|
|
|||||
100-44-20UG
|
Gibco™
1004420UG |
20 μg |
Each for $211.00
|
|
|||||
10044250UG
|
Gibco™
10044250UG |
250 μg |
Each for $1,134.00
|
|
|||||
10044500UG
|
Gibco™
10044500UG |
500 μg |
Each for $1,944.00
|
|
|||||
100-44-5UG
|
Gibco™
100445UG |
5 μg |
Each for $87.00
|
|
|||||
Description
Endocrine gland-derived vascular endothelial growth factor induces proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. Its expression is induced by hypoxia and is restricted to the steroidogenic glands. Its expression is often complementary to the expression of VEGF, suggesting that these molecules function in a coordinated manner.Specifications
P58294 | |
protein with no preservative | |
9.6 | |
E. coli | |
RUO | |
black mamba toxin-related protein; EGVEGF; EG-VEGF; endocrine-gland-derived vascular endothelial growth factor; Mambakine; PK1; PRK1; PROK1; prokineticin 1; prokineticin-1; UNQ600/PRO1186 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
84432 | |
Human EG-VEGF | |
-20°C | |
<1 EU/μg | |
PROK1 | |
Unconjugated | |
AVITGACERD VQCGAGTCCA ISLWLRGLRM CTPLGREGEE CHPGSHKVPF FRKRKHHTCP CLPNLLCSRF PDGRYRCSMD LKNINF | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title