Learn More
Proteintech Group Inc Human CCL5 ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0009396T
Encompass_Preferred
CCL5, also known as RANTES, belongs to the CC chemokine family. It is strongly expressed in the human and murine liver upon injury. CCL5 has been found to be produced by numerous cell types, including platelets, macrophages, endothelial, and stellate cells. CCL5 recruits T cells, dendritic cells, eosinophils, NK cells, mast cells, and basophils to sites of inflammation by interacting with 3 specific G protein-coupled receptors: CCR1, CCR3, and CCR5. While CCL5 may promote efficient anti-tumour immune responses, it has also been associated with cancer progression and metastasis.
Catalog No. 50-197-4056
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.