Learn More
CELLULAR ENGINEERING TECHNOLOGIES INC Cryopreserved vial of human bone marrow-derived mesenchymal stem cells (500,000 cells)
Supplier: CELLULAR ENGINEERING TECHNOLOGIES INC CR1005500
Cells isolated from red marrow were collected from a single adult human donor and then passaged three times to assure consistency. Vial contains approximately 500,000 cells. Shipped with dry ice.Bone marrow-derived mesenchymal cells (BMSCs) contribute to the homeostasis of musculoskeletal tissue and support the growth and differentiation of primitive hemopoietic cells. BMSCs are incredibly versatile and have been used in various tissue differentiation applications, including but not limited to adipogenic, chondrogenic, osteogenic, and neural cell-like conversions. These cells can be used for multiple tissue engineering and regenerative medicine applications.Vial contains approximately 500,000 cells. Shipped with dry ice.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.