Learn More
Invitrogen™ Human beta Amyloid (1-42) PTD Recombinant Protein
Major constituent of plaques and tangles that occur in Alzheimer's disease (AD) patients.
$333.00 - $600.00
Specifications
For Use With (Application) | Assay Standard |
---|---|
Gene ID (Entrez) | 351 |
Species | Human |
Content And Storage | Store in freezer (-5 to -30°C). |
Shipping Condition | Dry Ice |
Description
Human beta Amyloid (1-42) PTD Recombinant Protein for Western Blot, IHC, ELISA, Ctrl
Format/Formulation: Lyophilized trifluoroacetate salt
Molecular Weight: 4515
Specifications
Assay Standard | |
Human | |
Dry Ice | |
Neurobiology |
351 | |
Store in freezer (-5 to -30°C). | |
Amyloid-Related Proteins, Neuropeptides |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.