Learn More
ACROBiosystems ACROBiosystems Human ANGPT2-coupled Magnetic Beads
Supplier:  ACROBiosystems MBSK0352MG
Encompass_Preferred
Beads;2MG;The magnetic beads technology makes use of the easy and efficient collection of beads in magnetic field to facilitate antibody purification in a simple workflow of “bind-wash-elute”. In contrast to common separation techniques, this method does not require columns or centrifugation, and is therefore ideal in high-throughput applications. ;Immobilized 47μg ANGPT2 protein/1mg beads can bind Human TIE2 Protein, Fc Tag (Cat.No. TI2-H5255) with an EC50 of 3.228 μg/mL (QC tested).
Catalog No. 50-210-6313
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.