Learn More
Bethyl Laboratories, Inc Human Albumin Elisa Kit
SDP
Supplier:  Bethyl Laboratories, Inc E88129
Encompass_Preferred
Human Albumin ELISA Kit, 1 xpre-coated 96-well strip plate, standard, detection antibody, sample dilution buffer, HRP solution, TMB substrate, stop solution, 20X wash buffer, sealing tape
Catalog No. NC9015905
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.