Learn More
ACROBiosystems Human CD3E & CD3D Heterodimer-coupled Magnetic Beads
Supplier:  ACROBiosystems MBSK00310MG
Encompass_Preferred
Human CD3E & CD3D Heterodimer protein is expressed from human 293 cells (HEK293).It contains AA Asp 23 - Asp 126 (CD3E) & Phe 22 - Ala 105 (CD3D) (Accession # P07766-1(CD3E) & P04234-1(CD3D)).
Catalog No. 50-233-1823
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.