Learn More
Bethyl Laboratories, Inc Human Hemoglobin F Elisa Set
SDP
Supplier:  Bethyl Laboratories, Inc E80136
Encompass_Preferred
Human Hemoglobin F ELISA Quantitation Set, 1000 wells, Set Components Affinity purified coating antibody, calibrator for standard curve, HRP detection antibody,Antigen:Hemoglobin F,Antigen Species: Human,Species Reactivity:Human
Catalog No. NC0188657
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.