Learn More
Wilmad Labglass HT MNTL KTTL AL HOUSING 4000ML
Supplier:  Wilmad Labglass LG8883110
Encompass_Preferred
Heating Mantle, Reaction Kettle, Aluminum Housing. Size: 4000mL; Watts: 750. Volts: 115. Aluminum housing exterior-designed to be used w/ reaction vessels LG-8071- through LG-8082. Supplied w/ 3-Wire cord and locking connector.
Catalog No. 50-801-232
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.